Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2xYT Agar (Powder/Molecular Genetics), Fisher BioReagents
Specifications
Additive | 2xyt Agar |
---|---|
Form | Powder |
For Use With (Application) | Molecular Genetics |
Description
Specifications
2xyt Agar | |
Powder | |
Molecular Genetics |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title