Learn More
World Precision Instrument 2mm Shielded H2S Sensor
SDP
Supplier:  World Precision Instrument ISOH2S2
Encompass_Preferred
About Hydrogen Sulfide
Although hydrogen sulfide is generally thought of in terms of a poisonous gas, it is endogenously produced in many mammalian tissues. It has been detected in micromolar amounts in blood and brain tissue. Hydrogen sulfide is reported as having a broad range of biological functions and although its potential to participate in cell signaling is clear, this biological role is not well understood. Hydrogen sulfide is strongly anagolous to nitric oxide because they share several physical and metabolic properties.
Catalog No. 50-252-1169