Learn More
Sarstedt Inc Screw cap micro tubes, 2 ml, PCR Performance Tested, Low protein-binding, 1000/pack
Supplier: Sarstedt Inc 72.694.600
2mL Screw Cap Micro Tube Protein Low Binding features knurls, assembled cap, skirted base, writing space and graduation.
This item is not returnable.
View return policy