Learn More
GVS™ 2mL Ampoules (50 Pack)
Ready-to-use 2mL ampoule is a liquid culture growth media that can save time and money by shortening prep time and minimizing possible contamination. The 2mL ampoules contain enough media to saturate a standard absorbent pad in a petri dish
$128.00 - $404.00
Specifications
Certifications/Compliance | Batch-specific quality certificate in each pack |
---|---|
No. per Pack | 50 |
Packaging Type | Ampoules |
Quantity | 2 mL |
Catalog Number | Mfr. No. | Broth Type | Detects | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Broth Type | Detects | Price | Quantity & Availability | ||||
10-486-138
|
GVS™
10496101 |
M-Green Yeast and Mold Broth | yeast and mold |
Pack of 50 for $128.00
|
|
10-486-139
|
GVS™
10496102 |
M-TGE Total Count Liquid Media | Bacteria |
Pack of 50 for $128.00
|
|
10-486-140
|
GVS™
10496103 |
M-Endo Coliform Broth | coliform |
Pack of 50 for $128.00
|
|
10-486-141
|
GVS™
10496104 |
Orange Serum Liquid Media | aciduric microorganisms |
Pack of 50 for $144.00
|
|
10-486-142
|
GVS™
10496106 |
PRY Broth | spoilage microorganism in beverages and water |
Pack of 50 for $128.00
|
|
10-486-145
|
GVS™
10496112 |
MRS Broth | supports luxuriant growth of all lactobacilli, even the slow growing species. |
Pack of 50 for $404.00
|
|
10-486-146
|
GVS™
10496113 |
Total Count Broth with TTC | Bacteria |
Pack of 50 for $128.00
|
|
10-486-147
|
GVS™
10496114 |
M-FC Broth with Rosolic Acid | allows the development of fecal coliforms at elevated temperatures (44.5°C) |
Pack of 50 for $128.00
|
|
10-486-148
|
GVS™
10496116 |
M-Green Select Broth | detection and enumeration of fungi, yeast and mold. |
Pack of 50 for $158.00
|
|
10-486-149
|
GVS™
10496119 |
Pseudomonas Broth | Pseudomonas Aeruginosa |
Pack of 50 for $128.00
|
|
10-486-150
|
GVS™
10496120 |
Enterococcus Broth | Direct count of enterococci in 48 hours. |
Pack of 50 for $181.00
|
|
10-486-151
|
GVS™
10496121 |
Mannitol Salt Broth | presumptive pathogenic Staphylococci. |
Pack of 50 for $181.00
|
|
10-486-152
|
GVS™
10496124 |
M-FC Broth | allows the development of fecal coliforms at elevated temperatures (44.5°C) |
Pack of 50 for $128.00
|
|
10-486-153
|
GVS™
10496125 |
KF-Streptococcus Broth | fecal streptococci in polluted surface waters. |
Pack of 50 for $128.00
|
|
10-486-154
|
GVS™
10496146 |
Cetrimide Broth | for selective cultivation of Pseudomonas aeruginosa |
Pack of 50 for $181.00
|
|
10-486-155
|
GVS™
10496151 |
Heterotrophic Plate Count (HPC) Broth with TTC | live heterotrophs in drinking water and other media |
Pack of 50 for $158.00
|
|
10-486-157
|
GVS™
10496164 |
Heterotrophic Plate Count (HPC) Broth | live heterotrophs in drinking water and other media |
Pack of 50 for $128.00
|
|
10-486-158
|
GVS™
10496192 |
MI-Broth Liquid Media | presence of coliform bacteria |
Pack of 50 for $276.00
|
|
Description
Features and Benefits
- Pre-Sterilized and Ready to Use
- Time Saving, 2 mL Portions, Reduce Risk of Cross-Contamination.
- 2mL ampoules contain enough media to saturate a standard absorbent pad in a petri dish
- Optimal media stability, sterility, and reproducibility
- Less time-consuming, higher productivity
- Batch-specific quality certificate in each pack, Confirms Reproducibility and Reliability
Applications
- Microbiological analysis.
- Detection of fungi in routine analysis of beverages using membrane filter technique.
- For testing soft drinks and other beverages for growth of yeast and mold
Liquid Media
Ready-to-use media considerably reduce the preparation time in quality control laboratories and also effectively reduce the risks of cross contamination. GVS is cooperating closely with quality assurance managers in the industry in the development of its own media and test kits. This intensive product development has produced a range of products that is being used to monitor production plants and conduct Microbiological checks on raw materials through to final product release in laboratories in more than 40 countries.
See 'GVS Life Sciences Microbiology Testing mpg YouTube' Video for further details on use.
Specifications
Batch-specific quality certificate in each pack | |
50 | |
Ampoules | |
2 mL |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.