Learn More
Ace Glass, Inc. 2L EXTRACTOR CHAMBER 2L Extractor Chamber
Supplier: Ace Glass, Inc. 684023
2L outer extraction chamber designed for the extracting of liquids with ether. The dispersion of the ether is accomplished with the use of a 70-100micron porosity ACE fritted disc, which is attached to the base of the funnel tube.