Learn More
United Scientific™ 2D Tubes with Push Caps
![SDP](/content/dam/fishersci/glyphs/sdp.png)
![SDP](/content/dam/fishersci/glyphs/sdp.png)
![SCI_ED](/content/dam/fishersci/glyphs/scied.png)
Tubes with push caps and 2D barcode include internal spiral design for secure air travel.
Supplier: United Scientific™ UNSLT0022DPCPK10
Description
2D Tubes with Push Caps are made of virgin polypropylene materials for the highest level of safety against heavy metal ions and hazardous materials. Their internal thread provides optimal sealing for storage at ultralow temperatures, and silicone seals allow for secure air travel. The one-piece process allows the code to be integrated into the tube body.
The 1 mL tubes are available with a unique 2D Data-Matrix code. Due to the in-mold labeling technique used to manufacture the sample storage tubes, the white code surface cannot be separated from the transparent tube.
- Code is firm and non-shedding, resistant to acid, alkali, organic solvents and mechanical friction
- Produced in ISO Level 8 (Class 100,000 cleanroom) and automated production line to ensure DNase/RNase- and pyrogen-free
- Original coding, digital co-distribution technology and pure black and white ultra-high contrast Data-Matrix code ensure reliability of code reading
- Tubes with internal thread can be sealed with 96-format TPE caps
- Transportation safety in accordance with IATA requirements
Specifications
Push Cap | |
Not Autoclavable | |
Flat | |
Internal | |
960 | |
Sterile |
ISO13485:2016 and EU CE Certification (IVDD Directive) | |
1 mL | |
Polypropylene (Medical Grade) | |
2D Barcode 96-well Rack | |
-196°C to 121°C | |
No |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.