Learn More
Cole-Parmer Partlow 54422011 10" Circular Chart Recorder; 2 Channel/2 Relay per Channel
Supplier:  Cole-Parmer 4900185
Encompass_Preferred
These recorders are designed with the latest innovation in recording technology, enclosures, and functionality. Finding a place to install this recorder is easy with the compact 2.5" panel depth and short 1.3" protrusion — cutout size is the commonly-utilized 12.7" square, too! It can be panel or surface mounted.
Catalog No. 50-628-398