Learn More
Ace Glass, Inc. 290mm H quartz immersion well, Ace-Thred connection
Supplier: Ace Glass, Inc. 785428
290mm H double-walled quartz immersion well modified with a 4.8mm I.D. PTFE tube attached to a glass tube, and secured with a #7 Ace-Thred bushing. Inlet tube extends down the annular space to insure flow of coolant from bottom of well upward to outlet. Well has 60/40 inner joint. O.D. of outer jacket is 53mm max. Length above joint is 76mm.