Learn More
High Purity Standards 26 ELEMENT STD 100UG/ML 250ML
Supplier: High Purity Standards QCS26R250
26 element ICP quality control standard: Al, Sb, As, B, Ba, Be, Cd, Ca, Co, Cr, Cu, Fe, Mg, Mn, Mo, Ni, Pb, K, Se, Si, Ag, Na, Tl, Titanium, V, Zn @ 100 ug/mL in 5% HNO3 + Tr HF