Learn More
Chemglass Life Sciences 25mL Glas-Col Micro Heating Mantle, Spherical, 30 Watts, 60 Volts
Supplier: Chemglass Life Sciences CG1400004
25mL Glas-Col Micro Heating Mantle, Spherical, 30 Watts, 60 Volts. The Glas-Col micro mantle provides precise heat control for small spherical flasks. This mantle is used extensively in analytical work and in purification of research products when only small samples are available. Supplied complete with a 3 foot, 2-wire cord and locking connector. NOTE: Due to the voltage of the micro mantle, a variable voltage transformer is required.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.