Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHOWA™ 257 Aramid Knit Cut-Resistant Glove Gloves, Foam Nitrile Coating
Click to view available options
Size:
6, Small
7, Medium
8, Large
9, X-Large
10, 2X-Large
Description
- 13-gauge plated-knit spandex liner with aramid and stainless steel
- Foamed flat dipped coating with sponge finish for secure grip
- Contact Heat level 2
- Lightweight, flexible liner with elastic knitted wrist
- Seamless knit and breathable open back design
- Soft and flexible glove engineered with stainless steel and aramid to withstand the highest level of cuts and lacerations
- Foam nitrile coating protects palm and fingers from abrasions, snags and punctures, while offering optimum grip in both dry and oily applications
- Plated-knit liner avoids scratchy fibres touching the skin, ensuring a comfortable glove for long lasting wear
- Lightweight, breathable liner with open back design reduces sweat and keeps hands dry
- Ergonomically designed to replicate the hand’s natural curves
- Launderable for re-use, leads to less waste and cost efficiency
Specifications
Certifications/Compliance | ANSI/ISEA 105-16: Level A7 (minimum 4000g) |
Certifications/Compliance (Europe) | EN 388:2016: Cut Level F |
Coating Material | Sponge Nitrile |
Color | Yellow, Black |
Cut Resistance Rating | ANSI A7 |
Material | Aramid Fiber, Spandex™, Stainless Steel |
Product Type | Cut Resistant Gloves |
Size | 6, Small |
EN388 Standard | EN 388:4X42F |
For Use With (Application) | Automotive, mechanical, engineering, metal stamping, glass, window manufacturing, component assembly |
Show More |