Learn More
Waters Corp 2555 Quaternary Gradient Module Performance Maintenance Kit
Supplier: Waters Corp 201000210
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The Waters Performance Maintenance Kit for the 2555 Quaternary Gradient Module contains all the Waters Quality Parts necessary to keep your instrument operating at peak performance on an annual basis. The Waters Performance Maintenance Kit contains the following parts: solvent filters, diffuser filters, check valve cartridges, plunger seals, o-rings, sapphire plungers, seals for plunger wash, oultet filter and vent valve rotor seal.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.