Learn More
Ace Glass, Inc. Fabric heating mantle that covers top and bottom of 1, 2 or3 neck 250mL spherical flask. Lower half wattage is 180w at115 volts, CSA rated
SDP
Supplier:  Ace Glass, Inc. 1203113
Encompass
Catalog No. 50-977-258
Due to product restrictions, please to purchase or view availability for this product.