Learn More
Chemglass Life Sciences Funnel, 250mL, Separatory, Blank, Stopper Neck, Size # 22, Glassblowers
Supplier: Chemglass Life Sciences CG62404
Glassblowers Friedrich Condenser Blank. Molded inner cold finger for use in fabrication of Friedrich style condensers. Overall length is 230mm with an outside diameter of 45mm. NOTE: TOP IS UNFINISHED.