Learn More
Chemglass Life Sciences 250mL Glassblowers RBF Blank
Supplier: Chemglass Life Sciences CG61806
250mL Glassblowers RBF Blank. Blanks for fabrication of single or multi-neck round bottom flasks. NECK IS UNFINISHED, FLASK CANNOT BE USED WITHOUT MODIFICATION OF NECK.