Learn More
Chemglass Life Sciences 250mL Peptide Synthesis Vessel, Solid Phase, 2mm T-Bore PTFE Stpk, Medium Frit, GL 32 Thread
SDP
Supplier:  Chemglass Life Sciences CG186405
Encompass_Preferred
250mL Peptide Synthesis Vessel, Solid Phase, 2mm T-Bore PTFE Stpk, Medium Frit, GL 32 Thread. Solid phase peptide synthesis vessel having a medium porosity, fritted glass resin support. Top of the vessel has a GL thread and is supplied complete with a PTFE lined PBT screw cap. Lower stopcock is a “T”-Bore 2mm PTFE to allow nitrogen to be used to upwardly agitate the resin beads while shaking. The O.D. of both lower stems is 8mm. Vessels are available in sizes other than those listed. Please call for details.
Catalog No. CG186405