Learn More
Chemglass Life Sciences 250mL Addition Funnel, Graduated, 29/26 Joint Size, 2mm PTFE Metering Stopcock, 350mm OAH
Supplier: Chemglass Life Sciences CG171316
250mL Addition Funnel, Graduated, 29/26 Joint Size, 2mm PTFE Metering Stopcock, 350mm OAH. Cylindrical style addition funnel with a PTFE adjustable bore metering plug. Turning the adjustment knob allows flow rate to be controlled from dropwise to full flow. For replacement stoppers see CG-3000.