Learn More
Chemglass Life Sciences Glas-col Mantle Extension Support for Series inMin, Mantle Size 250mL-500mL, 162mm ID
Supplier: Chemglass Life Sciences CG1100402
Glas-col Mantle Extension Support for Series inMin, Mantle Size 250mL-500mL, 162mm ID. Glas-Col extension support for use with a smaller Series M mantle. Attached to a stable ring stand or distillation rack, the support can be adjusted to any convenient height. Support is painted gray and has a 6in long, 1/2in O.D. rod.