Learn More
Cole-Parmer Argos Technologies PolarSafe Cardboard Freezer Box Divider, 4-7/8" x 4-7/8" x 1-5/16"; 25-Place
Supplier:  Cole-Parmer 0675581
Encompass_Preferred
Create customized storage box configurations with these freezer boxes. A snug-fitting lid prevents accidental separation if dropped. The boxes can be used in mechanical freezers or liquid nitrogen tanks up to 196C. They are crafted of sturdy cardboard with moisture-repellent coating and designed with small holes on the bottom to allow for drainage. To customize storage needs, cardboard freezer boxes can be purchased separately.
Catalog No. NC1480432