Learn More
Ace Glass, Inc. Tube only, pressure, 15mL, #15, 25.4mm OD, 10.2cm long, no plug
SDP
Supplier:  Ace Glass, Inc. 864823
Encompass_Preferred
15mL pressure tube with #15 Ace-Thred, rated for 150psig at as much as 120C. Does not include plug. Dimensions: 25mm O.D. x 10.2cm long.
Catalog No. 50-955-8502