Learn More
AGA Marvel 24-IN GENERAL PURPOSE ALL FREEZER
Supplier:  AGA Marvel MS24FAS2RW
Encompass_Preferred
Accurate electronic temperature control of contents within +2/ -2C of set point. Recommended ambient temperature 18.3C to 32.2C (65F to 90F) Large, easy-to-read digital temperature display with resolution to 0.1C User selectable temperature set point between -12C and -25C, factory preset to -20C User selectable high and low temperature alarm values User selectable alarm delay time period Visual and audible alarms for over temperature, under temperature, and door ajar events Alarm silence and automatic ring back functions Keypad lock-out to prevent unintended control inputs Basic high and low temperature logging with reset capability Visual diagnostic error codes
Catalog No. NC1232745