Learn More
Tripp Lite 2400 WATT 6OUTLET LINE COND.
Supplier: Tripp Lite LC2400
Line Conditioner/AVR System; Tripp Lite; 2400 watt; 6 outlets; Cord Length: 6 ft.; 20 amp capacity; Maintains regulated 120V nominal output over an input range of 89 to 147V