Learn More
Molecular Dimensions 24-WELL XRL PLATE 100/CS
Supplier: Molecular Dimensions MD311
Protein Crystallography. 24-well XRL Plate (100) This Linbro-style vapour diffusion crystal growth plate with raised lid has superior optical properties. The wells take 22mm cover slips for hanging and sitting drop.