Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ 24 Listeria Enrichment Broth (24 LEB) (Dehydrated)

Selectively enrich Listeria monocytogenes and other Listeria spp. from food and environmental samples.

Supplier:  Thermo Scientific™ CM1107B

Catalog No. CM1107B


Add to Cart
This item is not returnable. View return policy

Description

Description

Selectively enrich Listeria monocytogenes and other Listeria spp. from food and environmental samples with Thermo Scientific™ Oxoid™ 24 Listeria Enrichment Broth (24 LEB) (Dehydrated). The medium provides optimal growing conditions for all Listeria species for PCR applications. It is specifically designed for use with PCR systems, such as Thermo Scientific™ SureTect™ molecular assays for Listeria.

Listeria monocytogenes is the most common pathogenic Listeria spp. and has been shown to be pathogenic to both humans and animals. 24 Listeria Enrichment Broth (24 LEB) is designed to identify Listeria spp. based on their utilization of a chomogenic substrate.
  • Easy-to-read: chromogenic and phospholipase enzyme reactions easily differentiate Listeria monocytogenes from other Listeria spp.
  • Adheres to ISO 11290-1:1997 standards.
  • Superior to PALCAM and Oxford medium for the isolation of Listeria monocytogenes1.
The presence of the chromogenic substrate for the detection of  glucosidase (an enzyme common to all Listeria spp. and to a few strains of Enterococci and Bacilli) results in chromogenic activity.
Further differentiation of Listeria monocytogenes and other Listeria spp. is facilitated by the addition of phosphatidylinositol or lethicin substrate for the detection of phospholipase enzyme.

Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

10°C to 30°C
Powder
500 g
24 Listeria Enrichment Broth (24 LEB) (Dehydrated)
Dehydrated Culture Media
Product Suggestions

Product Suggestions

Safety and Handling

Safety and Handling

ShelfLife : 5 Years

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit