Learn More
Streck CVA™ Calibration Verification Assessment/Linearity Kit
CVA (Calibration Verification Assessment) and CVA for CELL-DYN are assayed linearity control kits that test the upper and lower limits of three- and five-part differential hematology analyzers to determine their patient reportable range and linear performance.
Supplier: Streck 291709
This item is not returnable.
View return policy
Description
- Provides assay values for the most popular three- and five-part hematology instruments.
- Customized kits with WBC, RBC, Hgb and platelet ranges for each instrument model.
- Plastic vials with pierceable caps for analyzer autosampling.
- 5-day open-vial stability, 120-day closed-vial stability.
- Tests the extreme upper and lower limits of an instrument.
- Access to STATS™, Streck’s free interlaboratory quality control program for peer group data comparison.
Specifications
AcT 5diff, HmX | |
WBC, platelet and RBC or Hemoglobin vials | |
5 | |
5 Days Open, 120 Days Closed Vial |
Linearity Control Kit | |
CVA for WBC, Platelets, HgB | |
12 x 3 mL |