Learn More
Justrite™ 23 Gallon ChemCor™ Under Fume Hood Corrosives/Acids Safety Cabinet With Self-Closing Doors
Increase workplace safety and improve cleaniliness with the proper safety and storage equipment. The Justrite™ 23 Gallon ChemCor™ Under Fume Hood Corrosives/Acids Safety Cabinet With Self-Closing Doors provides superior corrosion protection as well as fire protection in a single cabinet.
$5970.00
Specifications
Door Style Self-close
Load Bearing Capacity 23 gal.
View More Specs
Products 3
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
19-822-374
View Documents
Justrite™
8837222
Blue
Each for $5,970.00
Add to cart
19-822-375
View Documents
Justrite™
8837272
Light Neutral
Each for $5,970.00
Add to cart
19-822-376
View Documents
Justrite™
8837242
Silver
Each for $5,970.00
Add to cart
Description
Description
  • Expand storage, add new convenience to existing cabinets, or customize your new cabinet with additional shelves to meet specific workflow needs
  • Galvanized steel shelves hold up to 250 lbs and adjust on 3 in. increments for versatile storage
  • Patented SpillSlope galvanized steel shelves direct spills to back and bottom of a leakproof sump
  • Polyethylene accessories with a slightly curved edge provide additional protection against leaks and spills
Specifications
Specifications
Self-close
23 gal.