Learn More
Justrite™ 23 Gallon ChemCor™ Under Fume Hood Corrosives/Acids Safety Cabinet With Manual-Closing Doors
Increase workplace safety and improve cleaniliness with the proper safety and storage equipment. The Justrite™ 23 Gallon ChemCor™ Under Fume Hood Corrosives/Acids Safety Cabinet With Manual-Closing Doors provides superior corrosion protection as well as fire protection in a single cabinet.
$5190.00
Specifications
Door Style Manual-close
Load Bearing Capacity 23 gal.
View More Specs
Products 3
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
19-822-372
View Documents
Justrite™
8837072
Light Neutral
Each for $5,190.00
Add to cart
19-822-373
View Documents
Justrite™
8837042
Silver
Each for $5,190.00
Add to cart
19-822-371
View Documents
Justrite™
8837022
Blue
Each for $5,190.00
Add to cart
Description
Description
  • Expand storage, add new convenience to existing cabinets, or customize your new cabinet with additional shelves to meet specific workflow needs
  • Galvanized steel shelves hold up to 250 lbs and adjust on 3 in. increments for versatile storage
  • Patented SpillSlope galvanized steel shelves direct spills to back and bottom of a leakproof sump
Specifications
Specifications
Manual-close
23 gal.