Learn More
Bellco Glass, Inc. 2.2mL Bacteriological/Milk
Supplier: Bellco Glass, Inc. 229268000
All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets.Specifications Bacteriological/Milk pipets are manufactured to meet APHA requirements Specialized graduations allow quick volume identification Color Code Purple 50 Individually Wrapped Case Quantity 250