Learn More
Ace Glass, Inc. Aluminum housed Stirmantle for 22 liter spherical flask. Variable speed stirring is independent of heating making this unit Ideal for continuous distillations. Ships complete with motor, stir control, stir bar and cord, 120v 50/60Hz
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Ace Glass, Inc. 1204522
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Aluminum housed Stirmantle for 22L spherical flasks by Glas-Col. Mantle is IDeal for continuous distillations by suppling controlled heating and stirring in one unit. Ships complete with motor, stir control, stir bar, and four foot cord. 120v 50/60Hz and 100-1850rpm.