Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

American Educational Products 21st Century New York Balance
SCI_ED

A new era in balances.

Supplier:  American Educational Products 71451

Catalog No. S66414

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Includes:
  • 20 in. (50cm) stick
  • Three knife-edge clamps
  • 50 and 100g hooked weights

Description

Description

Demonstration helps students understand the relationship between length of the lever arm, moment of force, and amount of force. Adjusting the weights on the level shows the law of torque and its properties.

  • 1.7 in. (0.5m) stick, graduated in centimeters
  • Lever clamp that fits on the stick to form the fulcrum for the lever
  • 50g and 100g knife-edge masses
  • Support base for lever clamp

Order Info

Dimensions: 19.5 x 3.8 x 6.5 in. (49 x 9.4 x 16.25cm); Weight: 1.6 lb.

Specifications

Specifications

21St Century NY Balance
20 in. Stick, three knife edge clamps, 50 and 100g hooked weight
2.2 kg
To demonstrate the students
5 lb.
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit