Learn More
Chemglass Life Sciences 20L UnJacketed Cylindrical Process Filter Reactor System Complete, Air Motor
Supplier: Chemglass Life Sciences CG1958F821
20L UnJacketed Cylindrical Process Filter Reactor System Complete, Air Motor. Chemglass's unjacketed filter reactors can be used for single or multi-step reactions and filtrations in the same vessel, production and process scale peptide synthesis. Filter base eliminates material transfers and significantly reduces losses. Complete reactor system on Chemglass's open air, autocenter stand. Please Note: Filter Reactors are for Vacuum/Gravity Filtration Use Only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.