Learn More
Sino Biological SARS-CoV-2 (2019-nCoV) Spike S1 Gene ORF cDNA clone expression plasmid (Codon Optimized) 1Unit
Supplier: Sino Biological VG40591UT
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
A number of silent mutations were introduced into the DNA sequence in order to increase its protein expression level in mammalian cell system.The translated amino acid sequence is identical with QHD43416.