Learn More
LabScientific 200UL, 0.2 mL (9.5 mm max) CARDBOARD MICRO TUBE BOXES, 169 WELLS, 13x13 format, 10/PK
Supplier: LabScientific 220
200UL, 0.2 mL (9.5 mm max) CARDBOARD MICRO TUBE BOXES, 169 WELLS, 13x13 format, 10/PK. They are designed to ensure that the tubes contained will remain in their appointed well even if the box is tipped or knocked when the lid is in place. Will fit into most standard freezer racking.