Learn More
Chemglass Life Sciences Reaction Kettle Lid, Duran, Flat Ground, Lid ID: 200mm
Supplier: Chemglass Life Sciences CG14905
Reaction Kettle Lid, Duran, Flat Ground, Lid ID: 200mm. Dome style lid for construction of single or multiple neck reaction kettle heads. Mating surface is ground. Outer taper of flange is formed at a 45 angle to accept the CG-141 stainless steel clamps.