Learn More
Chemglass Life Sciences Reaction Kettle Flange, Duran, Flat Ground, Flange ID: 200mm
Supplier: Chemglass Life Sciences CG14805
Reaction Kettle Flange, Duran, Flat Ground, Flange ID: 200mm. Similar to CG-147, but formed without the o-ring groove.