Learn More
Chemglass Life Sciences 2000mL Glassblowers RBF Blank
SDP
Supplier:  Chemglass Life Sciences CG61811
Encompass_Preferred
2000mL Glassblowers RBF Blank. Blanks for fabrication of single or multi-neck round bottom flasks. NECK IS UNFINISHED, FLASK CANNOT BE USED WITHOUT MODIFICATION OF NECK.
Catalog No. CG61811