Learn More
Chemglass Life Sciences Glas-Col Mantle Extension Support for 2000mL Series O Mantle, 9 1/4in ID
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Chemglass Life Sciences CG1001006
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Glas-Col Mantle Extension Support for 2000mL Series O Mantle, 9 1/4in ID. This lattice mount support holds spherical and hemispherical Glas-Col Series O mantles up to 5-liter capacity. The steel support, with its fabric strap basket, is strong, handy and durable. Attaches easily to stable ring stands or distillation racks. Supports are painted gray. This mantle support is made from 3/8in diameter rod.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.