Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Troemner™ 20 g-100 mg Analytical Precision UltraClass Weight Set
Combine high precision with the advantage of two-piece construction. UltraClass weights, exclusive to Troemner™ are the most precise two-piece weights available with weight tolerances that equal ANSI/ASTM E617 Class 0 or OIML R 111 Class E2 tolerances.
$1592.00 - $2508.00
Specifications
Classification | UltraClass |
---|---|
Material | Troemner Alloy 8, Stainless Steel |
Case Material | Polycarbonate |
Density | Weights 1 g and above are 8.03 g/cm3 at 20°C. Weights below 1 g are 7.95 g/cm3 at 20°C. |
Mass | 20 g to 100 mg |
Catalog Number | Mfr. No. | Certifications/Compliance | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Certifications/Compliance | Price | Quantity & Availability | |||||
02-218-069
|
Troemner™
30390066 |
No Certificate |
Each for $1,592.00
|
|
|||||
02-218-070
|
Troemner™
30390103 |
Traceable Certificate |
Each for $1,811.00
|
|
|||||
02-218-071
|
Troemner™
30390140 |
NVLAP Accredited Certificate |
Each for $2,508.00
|
|
|||||
Description
- Used in extremely stable environments
- 1 to 500 mg weights are one-piece construction with no adjusting cavity with one end turned up for easy handling
- Weights 1 g and above are of two-piece construction
- Weight sets 1 mg-500 g are provided in a high-quality polycarbonate set case
Specifications
UltraClass | |
Polycarbonate | |
20 g to 100 mg | |
8.89 cm | |
Calibration of high-precision analytical balances with readability as low as 0.01 mg | |
10/Set |
Troemner Alloy 8, Stainless Steel | |
Weights 1 g and above are 8.03 g/cm3 at 20°C. Weights below 1 g are 7.95 g/cm3 at 20°C. | |
20.32 cm | |
Reference standard calibration | |
Cylindrical Weight: 20 g, 10 g, 5 g, (2) 2 g, 1 g, Sheet Weight: 500 mg, (2) 200 mg, 100 mg, Polycarbonate Case, Forceps, Brush |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title