Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Propanol for HPLC, 99.5%, MilliporeSigma™ Supelco™
Click to view available options
Quantity:
50 L
Packaging:
PVC-coated Bottle
Description
2-Propanol (i-PrOH), a renewable secondary alcohol, is water soluble, colorless, volatile liquid with a sweet odor. It is a potential substitute to petroleum-derived gasoline. Preparation of methyl isobutyl ketone (MIBK) using 2-propanol as the precursor has been reported with yield as high as 25%. Solubility of benzoic acid has been investigated in 2-propanol by gravimetric method in the temperature range, 277-346K. Its photocatalytic degradation has been examined by various methods.
Specifications
Melting Point | -89.5°C |
Density | 0.785 g/mL (at 25°C) |
Boiling Point | 82°C |
Quantity | 50 L |
Packaging | PVC-coated Bottle |
Refractive Index | n20/D 1.377 |
Linear Formula | (CH3)2 CHOH |
Vapor Pressure | 44 mmHg (25°C) |
Formula Weight | 60.10 |
Percent Purity | 99.5% |
Show More |
Safety and Handling
RTECSNumber | NT8050000 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction