Learn More
Sigma Aldrich Fine Chemicals Biosciences 2-Propanol, 67-63-0, MFCD00011674, 4X25mL
Supplier: Sigma Aldrich Fine Chemicals Biosciences I95164X25ML
Linear Formula: (CH3)2CHOH, Molecular Weight: 60.10, BioReagent, for molecular biology, ≥99.5%, Synonym: sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol. 2-Propanol has been used: in DNA and RNA extraction, for determination of hydroxyproline levels, for checking cell viability by MTT assay, and in microarray hybridization.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.