Learn More
Ready America™ 2 Person 3 Day Deluxe Dry Backpack
The 2-Person Deluxe Dry-Pack includes all the needed essentials for the first 72-hours after an emergency or disaster. And it's all packed into a premium tactical style backpack. You get emergency food and water, tools, first aid, hygiene, and more.
Supplier: Ready America™ 70471
Description
- First responders advise us that we should be prepared for at least the first 72 hours after a disaster, it is absolutely essential that we have emergency supplies at our disposal. That is why Ready America has developed the 2 Person Deluxe Dry-Pack with the essentials for 3 days. It contains all the life saving supplies and more needed to survive for those crucial 72 hours.
- The Emergency Power Station is a four-function hand-crank unit that includes a flashlight, AM/FM radio, cell phone charger and a siren. One minute of cranking produces either 30 minutes of bright light, 2 minutes of talk time on a cell phone, or 15 minutes of radio. The Heavy Duty Multi-Tool includes: knife, screwdriver, bottle opener, saw, fire starter and pliers.
- Emergency Kit Contains:
- (2) Food Bar (3600 Calorie) (5-year shelf life)
- (18) Pouches of Water (4.225 oz.) (5-year shelf life)
- (1) Potable Aqua Germicidal Tablets (50 count)
- (3) Bio Hazard Bags
- (4) 10 in. Nitrile Gloves
- (1) Heavy Duty Work Gloves
- (2) N-95 Approved Dust Mask
- (1) First-Aid Kit (107 piece)
- (6) Insect Repellent Wipes (.27 oz.)
- (1) Heavy Duty Multi-Tool
- (1) Duct Tape (10 yards)
- (1) 5 in 1 Survival Whistle
- (2) Emergency Poncho
- (2) Emergency Survival Blanket
- (2) Knit Cap (Beanie)
- (2) Pocket Tissue
- (12) Wet Naps
- (1) Micro Spark Wheel
- (1) Heritage Light-Me Tinder (12 pack)
- (1) Insta-Fire (3 pack)
- (1) Deck of Playing Cards
- (1) Emergency Power Station 4 Function
- (6) 12 Hour Light Stick
- (1) Water Bottle (1L)
- (1) Hygiene Kit Basic
- (1) Extra Toothbrush
- (1) Extra Mouthwash
- (1) Hand Sanitizer (4 oz.)
- (1) 25 ft. Cord
- (1) Document Pouch
- (1) Deluxe Dry-Bag Backpack (30 L)
Specifications
Emergency Backpack | |
Gray | |
Reusable |