Learn More
Cambridge Isotope Laboratories BENZOIC ACID CALIBRANT FOR QNMR 15mM BENZOIC ACID IN DMSO-D6 (160 UL FILL VOLUME), 1 3MM TUBE
Supplier: Cambridge Isotope Laboratories DLM7061B
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
BENZOIC ACID CALIBRANT FOR QNMR 15mM BENZOIC ACID IN DMSO-D6 (160 UL FILL VOLUME), 1 3MM TUBE