Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxyquinoline-4-carboxylic Acid 98.0+%, TCI America™

Catalog Number H101625G
Click to view available options
Quantity:
5 g
25 g
15733-89-8
C10H7NO3
189.17
MFCD00023942 MFCD00464512
MFSHNFBQNVGXJX-UHFFFAOYSA-N
2-hydroxyquinoline-4-carboxylic acid, 2-oxo-1,2-dihydroquinoline-4-carboxylic acid, 2-hydroxy-4-quinolincarboxylic acid, 2-hydroxycinchoninic acid, cinchoninic acid, 2-hydroxy, 4-carboxycarbostyril, 2-oxocinchoninic acid, 4-quinolinecarboxylic acid, 1,2-dihydro-2-oxo, 2-hydroxy-quinoline-4-carboxylic acid, 2-hydroxy-4-quinolinecarboxylic acid
85076
CHEBI:52045
2-oxo-1,2-dihydroquinoline-4-carboxylic acid
OC(=O)C1=CC(=O)NC2=CC=CC=C12
15733-89-8
C10H7NO3
189.17
MFCD00023942 MFCD00464512
MFSHNFBQNVGXJX-UHFFFAOYSA-N
2-hydroxyquinoline-4-carboxylic acid, 2-oxo-1,2-dihydroquinoline-4-carboxylic acid, 2-hydroxy-4-quinolincarboxylic acid, 2-hydroxycinchoninic acid, cinchoninic acid, 2-hydroxy, 4-carboxycarbostyril, 2-oxocinchoninic acid, 4-quinolinecarboxylic acid, 1,2-dihydro-2-oxo, 2-hydroxy-quinoline-4-carboxylic acid, 2-hydroxy-4-quinolinecarboxylic acid
85076
CHEBI:52045
2-oxo-1,2-dihydroquinoline-4-carboxylic acid
OC(=O)C1=CC(=O)NC2=CC=CC=C12
Color Yellow
Quantity 25 g
Formula Weight 189.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxyquinoline-4-carboxylic Acid
RTECSNumber GD3869000
TSCA No
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit