Learn More
AGRIFY CORPORATION Cascade Sciences CVO-5 DOUBLE UP Package
Supplier: AGRIFY CORPORATION CS1901010047
Best selling, turnkey vacuum purge package in a mobile rack includes: 2 each CVO-5 Vacuum Ovens (5 cubic ft each) Ambient +26°F to 302°F (+14.4°C to 150°C) / 10 Total Shelves / powerful, oil-free, clean, quiet vacuum pump system, 1 each Industrial Rack on Casters with 2 Pump Shelves / 2 each MAX EVAP Connection Kits and Double Oven To Pump Connection Kit, 120V, NRTL
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.