Learn More
Electron Microscopy Sciences 2 Counting Units, plus Non-Locking Totalizer
Supplier: Electron Microscopy Sciences 7260204
For a quick, accurate count, the durable, long-lasting, ergonomic and easy to use Denominator is the perfect solution for fast, accurate tallies. The counters may be banked or tiered, and various mounting options are available to suit your needs. Totalizer locks every 100 counts. A release button quickly unlocks for the next 100 counts. Percentages by dividing the counter readings by the number of hundreds counted.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.