Learn More
Eurofins Discoverx 2-Arachidonylglycerol, 10 mg. (92-1226)
Supplier:  Eurofins Discoverx 921226
Encompass_Preferred
2-Arachidonylglycerol is an endocannabinoid ester formed by arachidonic acid and glycerol that is expressed in the CNS at much higher levels than anandamide. Has neuromodulatory effects, and is a potent agonist of the GPR55, CB1, and CB2 receptors (EC50=3, 519, and 618 nM, respectively). Provided as 10 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9749