Learn More
2,4-Dichloropyrido[3,2-d]pyrimidine, 95%, Thermo Scientific™
Specifications
Melting Point | 175.0°C to 177.0°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 94% min. (HPLC) |
Quantity | 250 mg |
Formula Weight | 200.03 |
Percent Purity | 95% |
Chemical Name or Material | 2,4-Dichloropyrido[3,2-d]pyrimidine |
Safety and Handling
GHS H Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.