Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2,3,6,7-Tetrakis(2-cyanoethylthio)tetrathiafulvalene 98.0+%, TCI America™
Click to view available options
Quantity:
100 mg
1 g
Specifications
Melting Point | 191°C |
Quantity | 1 g |
UN Number | 3439 |
Formula Weight | 544.83 |
Percent Purity | ≥98.0% (HPLC,N) |
Physical Form | Crystalline Powder |
Chemical Name or Material | 2,3,6,7-Tetrakis(2-cyanoethylthio)tetrathiafulvalene |
Safety and Handling
TSCA : No