Learn More
CELLTREAT Scientific Products 2.2mL Bacteriological/Milk Pipet, Bulk Packed, Sterile pipette
Supplier: CELLTREAT Scientific Products 229258
CELLTREAT Scientific Products 2.2mL Milk Pipet. Bacteriological/Milk Pipets are manufactures to meet APHA requirements shown in Standard Methods for Examination of Dairy Products and ASTM E1043-00(2010). Specialized graduations allow quick volume identification. Premium material barrier plug. Color coded stripes and packaging enable quick volume identification. RNase/DNase Free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, bulk packed. 50 bulk packed/bag, 400/case.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.