Learn More
Ace Glass, Inc. 1L jacketed filter reactor body with 100mm ID grooved top flange, #80 bottom thread, 100mm IDx180mm depth, with CAPFE O-ring, #11 Ace-Thred inlet/outlet connections, use with 3/8in ID tubing, pressure rated at 35 psi.
Supplier: Ace Glass, Inc. 6386316
1L jacketed filter reactor body with 100mm ID grooved top flange, #80 bottom thread, 100mm IDx180mm depth, with CAPFE O-ring, #11 Ace-Thred inlet/outlet connections, use with 3/8in ID tubing, pressure rated at 35 psi.